REAGENT KIT FOR PRIMELAB ID64(250 TESTS) 1 * 1 items
Catalog # WTIDPL64-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID64(250 TESTS) 1 * 1 items
Catalog # WTIDPL64-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
EOSIN RAL 555 1L 1 * 1 L
Supplier: RAL
Acriflavine hydrochloride fluorescent dye, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Formalin 10% histological fixative, neutral buffered, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Immersion Oil, fluorescence-free for microscopy 1 * 100 mL
Supplier: PanReac AppliChem
Sodium hydroxide 32% in aqueous solution
Supplier: Roth Carl
Toluidine blue O, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Stemgent Stemolecule All-Trans Retinoic Acid (100mg) 1 * 100 mg
Supplier: MARKET SOURCE PART PROCESS
FORMLABS Resin Washing Solution (5 L) 1 * 1 L
Supplier: MARKET SOURCE PART PROCESS
Diluid III Diff for hematology, J.T.Baker®
Supplier: Avantor