REAGENT KIT FOR PRIMELAB ID37 (50 TESTS) 1 * 1 items
Catalog # WTIDPL37-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID37 (50 TESTS) 1 * 1 items
Catalog # WTIDPL37-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hydroxide 1 mol/l, EMPROVE® EXPERT, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Test strips, lactic acid, Reflectoquant®, Supelco®
Supplier: Merck
Test strips, total hardness, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Visual colour kits, nitrite, MQuant®, Supelco®
Supplier: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach