REAGENT KIT FOR PRIMELAB ID10(50 TESTS) 1 * 1 items
Catalog # WTIDPL10-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID10(50 TESTS) 1 * 1 items
Catalog # WTIDPL10-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Total Iron Test Strips, Dosatest, VWR®
Supplier: VWR Chemicals
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Visual colour kits, cyanide, MQuant®, Supelco®
Supplier: Merck
METHYLENE BLUE RAL 555 AQUEOUS SOL. 2.5L 1 * 2,5 L
Supplier: RAL
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond