SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
REAGENTS KIT FOR PRIMELAB ID147(39TESTS) 1 * 1 items
WTIDLGP-40
:
WATER ID PRIMELAB
REAGENTS KIT FOR PRIMELAB ID147(39TESTS) 1 * 1 items
WTIDLGP-40
:
WATER ID PRIMELAB
Frequently Bought Together
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
: Merck
Test strips, nitrate/nitrite, Dosatest
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Immersion oil for microscopy, Sigma-Aldrich®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test strips for free chlorine, MQuant®, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test strips, sulphite, MQuant®, Supelco®
: Merck