25 TUBETESTS COD HR (0 - 15000 MG/L) 1 * 25 Vial
Catalog # WTIDCOD-17-HR
Supplier: WATER ID PRIMELAB
25 TUBETESTS COD HR (0 - 15000 MG/L) 1 * 25 Vial
Catalog # WTIDCOD-17-HR
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Eosin Y ∼99% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Calcium hydroxide 95%
Supplier: Thermo Fisher Scientific
TriLink® CleanCap® Beta gal mRNA
Supplier: TriLink BioTechnologies
CUBE SMALP 200 10 X 50 MG 1 * 500 mg
Supplier: MARKET SOURCE PART PROCESS
50 TABLETS PHENOL N° 1 PHOTOMETER 1 * 50 items
Supplier: WATER ID PRIMELAB
ACTINOMYCETE ISOLATION AGAR DEHYDRATED C 1 * 500 g
Supplier: HIMEDIA
VWR® Powder Packs for Determination of Iron, Reagents for Photometers
Supplier: VWR Chemicals
Nitrite VHR L, 500 ml
Supplier: Lovibond