Volumetric flask, PMP, transparent, 10 ml, NS 10/19, PP-stopper 1 * 2 items
Catalog # BRND36170
Supplier: Brand
Volumetric flask, PMP, transparent, 10 ml, NS 10/19, PP-stopper 1 * 2 items
Catalog # BRND36170
Supplier: Brand
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Facial Tissues
Supplier: VWR Collection
Sodium dihydrogen phosphate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Orthophosphoric acid ≥99%, EMSURE® for analysis, Supelco®
Supplier: Merck
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
VWR®, Universal Filter Tips
Supplier: VWR Collection
VWR®, Pipette Tips, Standard
Supplier: VWR Collection
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: EPPENDORF
VWR® Reagent Reservoirs
Supplier: VWR Collection
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Supplier: EPPENDORF