{[3-(2-IODOETHOXY)PROPOXY]METHYL}BENZENE 1 * 1 g
Catalog # UKROCSMB02125614-1
Supplier: UKRORGSYNTHESIS
{[3-(2-IODOETHOXY)PROPOXY]METHYL}BENZENE 1 * 1 g
Catalog # UKROCSMB02125614-1
Supplier: UKRORGSYNTHESIS
Frequently Bought Together
SIGTECH Mek Solvent 5 L Kanister 1 * 5 L
Supplier: MARKET SOURCE PART PROCESS
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Diethyl ether EMPROVE ESSENTIAL Ph Eur,BP 1 * 190 L
Supplier: MERCK PRODUCTION CHEMICALS
Diethylene glycol monobutyl ether ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol ≥99.85%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Glycerol, 99% for synthesis 1 * 1.000 mL
Supplier: PanReac AppliChem
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Pyridine ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Lithium perchlorate 99+% for analysis
Supplier: Thermo Fisher Scientific