ContactUsLinkComponent
SHODEX GPC H-2004, PREPARATIVE SIZE EXCLUSION COLUMN, 20x500mm, 15µm, 1,500Å, CHLOROFORM 1 * 1 items
Catalog # SHOWF6102004
Supplier:  Resonac
SHODEX GPC H-2004, PREPARATIVE SIZE EXCLUSION COLUMN, 20x500mm, 15µm, 1,500Å, CHLOROFORM 1 * 1 items
Catalog # SHOWF6102004
Supplier:  Resonac
Frequently Bought Together