Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EQ ACETYLCHOLINE-D16 BROMIDE 1 * 0,05 g
Catalog # MSPPD-0380/0.05
Supplier:
MARKET SOURCE PART PROCESS
Expand image
EQ ACETYLCHOLINE-D16 BROMIDE 1 * 0,05 g
Catalog # MSPPD-0380/0.05
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Petroleum spirit, 100…140 °C, EMPLURA® (naphtha benzine), Supelco®
Supplier: Merck
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
Supplier: Merck
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Tetrachloroethylene 99%, extra pure
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99.0-101.5% ACS
Supplier: Thermo Fisher Scientific
Benzyl alcohol, extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene, extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific