Single shaft f. Transferpette S -8/12 5 - 50 µl 1 * 2 items
Katalognr. BRND705620
Supplier: Brand
Frequently Bought Together
MEDELA EINWEG SEKRETBEUTEL 2.5 LITER MIT GELIERGRANULAT 1 * 30 ST
Supplier: MARKET SOURCE PART PROCESS
Masterflex® Ismatec® Replacement Cartridges and Adapters, Avantor®
Supplier: Avantor Fluid Handling
Accessories for Ultrapure Water System, Barnstead
Supplier: Thermo Fisher Scientific
DIGITAL-POTI 12mm 1 * 1 items
Supplier: IKA
Gasket Manometer+Steam release cock packed 1 * 1 items
Supplier: CERTOCLAV
Conductor 1 * 1 items
Supplier: ELMA SCHMIDBAUER
PLUG WITH BARBED TUBE FITTING BLUE 1 * 1 items
Supplier: Integra
Accessories for Diaphragm Pumps
Supplier: KNF
Extension board 1 * 1 items
Supplier: VWR Collection
XonaChip® 450 µm barrier 5PK includes 2 ml tube XC Pre-Coat 1 box of 5 1 * 5 items
Supplier: MARKET SOURCE PART PROCESS
MAINTENANCE KIT FOR MD 4C NT 1 * 1 items
Supplier: VACUUBRAND
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
VALVE FOR AERATION LID 1 * 1 items
Supplier: Retsch GmbH
wire protection assembly 1 * 1 items
Supplier: Binder
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach