ContactUsLinkComponent
,,6 components: Cr 250mg/kg Cd 250mg/kg K 250mg/kg Pb 250mg/kg P 250mg/kg Hg 50mg/kg in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 50 mL
Catalog # CMPLRM084188L05
,,6 components: Cr 250mg/kg Cd 250mg/kg K 250mg/kg Pb 250mg/kg P 250mg/kg Hg 50mg/kg in HNO3 5% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 50 mL
Catalog # CMPLRM084188L05
Frequently Bought Together