10 COMPONENTS: 1000MG/L EACH 1 * 100 mL
Catalog # CMPLRM093091L1
Supplier: CUSTOM MADE CHEMICALS LAB
10 COMPONENTS: 1000MG/L EACH 1 * 100 mL
Catalog # CMPLRM093091L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Embedding and mounting media
Supplier: Merck
NANOCOLOR® Photometric Tube Test Kits for Nitrate
Supplier: MACHEREY-NAGEL
60G PL SILICA HR 2 (241 TESTS) 1 * 60 g
Supplier: WATER ID PRIMELAB
Haematoxylin solution (Mayer), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
40G PL SILICA LR 3 (190 TESTS) 1 * 40 g
Supplier: WATER ID PRIMELAB
SULPHURIC ACID 0.01M (0.02N) ISO 17034 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Avantor® Seradigm, Fetal Bovine Serum (FBS), US origin
Supplier: VWR Seradigm
Ammonia solution 28.0 - 30.0% (NH₃ basis) in water ACS, Fluka™
Supplier: Honeywell Chemicals
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
CYTEK CFLUOR™ VIADYE RED FIXABLE VIABILITY DYE 1 * 1 items
Supplier: CYTEK BIOSCIENCES
SILVER NITRATE STD SOLUTION 0.100N 1 * 1 L
Supplier: Hach
Potassium hydroxide 85.0-100.5%, pellets Ph. Eur.
Supplier: VWR Chemicals
Erythrosin B disodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BOND ASPIRATING PROBE CLEANING SYSTEM 1 * 1 KIT
Supplier: LEICA MICROSYSTEMS