ContactUsLinkComponent
1 component: Hg 0.01mol/l in HCl 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Catalog # CMPLRM0931521L
1 component: Hg 0.01mol/l in HCl 5% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Catalog # CMPLRM0931521L
Frequently Bought Together