0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Plasmid DNA Marker, 3.2 mL. 1 bp lower marker 100,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-940 k it. 1 * 3,2 mL
Catalog # AGLSFS-SMK940-0003
Supplier:
AGILENT
Expand image
Plasmid DNA Marker, 3.2 mL. 1 bp lower marker 100,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-940 k it. 1 * 3,2 mL
Catalog # AGLSFS-SMK940-0003
Supplier:
AGILENT
Frequently Bought Together
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cell Strainers
Supplier: VWR Collection
Culture dishes for <i>in vitro</i> fertilisation, Nunc™
Supplier: Thermo Fisher Scientific
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, PP
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection