ContactUsLinkComponent
1 kb Plus DNA Ladder, 100 µL. 200-10,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-930 kit or the ZAG DNA Analyzer system and the ZAG-130 kit. 1 * 0,1 mL
Catalog # AGLSFS-SLR930-U100
Supplier:  AGILENT
1 kb Plus DNA Ladder, 100 µL. 200-10,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-930 kit or the ZAG DNA Analyzer system and the ZAG-130 kit. 1 * 0,1 mL
Catalog # AGLSFS-SLR930-U100
Supplier:  AGILENT
Frequently Bought Together