FP 55 kb BAC Ladder, 35 µL. 75-48,500 bp DNA Ladder for use with the Femto Pulse system and the FP-1003 kit 1 * 35 µG
Catalog # AGLSFP-7003-U035
Supplier: AGILENT
FP 55 kb BAC Ladder, 35 µL. 75-48,500 bp DNA Ladder for use with the Femto Pulse system and the FP-1003 kit 1 * 35 µG
Catalog # AGLSFP-7003-U035
Supplier: AGILENT
Frequently Bought Together
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
Akura™ Tilting Stand 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Scoops, long handled, SteriWare®
Supplier: Sampling Systems
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One
Culture tubes, sterile, Cultubes™
Supplier: Simport Scientific
VWR®, Cell Culture Insert Plates, PET
Supplier: VWR Collection
2000mL Non-Treated Roller Bottle, Non-Vented Cap, Sterile 1 * 12 ST
Supplier: MARKET SOURCE PART PROCESS
MULTIP. BEAKER PP 120 ML ST 1 * 300 items
Supplier: Greiner Bio-One
Sample tubes
Supplier: SEMADENI
PCR tubes and caps, Axygen®
Supplier: Corning
VWR® Square Polycarbonate Media Bottles
Supplier: VWR Collection
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with/without screw caps
Supplier: Thermo Fisher Scientific
NEOBOX 81 PLUS, UNTERTEIL BLAU 1 * 1 ST
Supplier: NEOLAB
Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling