ContactUsLinkComponent
5x Capillary Conditioning Solution, 50mL. Solution necessary for conditioning and restoring capillary array performance. 5x concentr ation. For use with the Fragment Analyzer and ZAG DNA Analyzer systems. 1 * 50 mL
Katalognr. AGLSDNF-475-0050
Supplier:  AGILENT
5x Capillary Conditioning Solution, 50mL. Solution necessary for conditioning and restoring capillary array performance. 5x concentr ation. For use with the Fragment Analyzer and ZAG DNA Analyzer systems. 1 * 50 mL
Katalognr. AGLSDNF-475-0050
Supplier:  AGILENT