ContactUsLinkComponent
Large Fragment Diluent Marker, 2.4 mL. 1 bp lower marker 100,000 bp upper marker. For use with the Fragment Analyzer systems and th e DNF-492 kit. 1 * 2,4 mL
Catalog # AGLSDNF-380-0003
Supplier:  AGILENT
Large Fragment Diluent Marker, 2.4 mL. 1 bp lower marker 100,000 bp upper marker. For use with the Fragment Analyzer systems and th e DNF-492 kit. 1 * 2,4 mL
Catalog # AGLSDNF-380-0003
Supplier:  AGILENT
Frequently Bought Together