SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
HS Genomic DNA Ladder, 100uL. 75-20,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-488 kit 1 * 100 µG
AGLSDNF-377-U100
:
AGILENT
HS Genomic DNA Ladder, 100uL. 75-20,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-488 kit 1 * 100 µG
AGLSDNF-377-U100
:
AGILENT
Frequently Bought Together
VWR®, Sample Containers, with Attached Lids
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Multiwell cell culture plates, Falcon®
: Corning
Transfer pipettes
: RATIOLAB
Pasteur pipettes, disposable
: Brand
Centrifuge tubes, conical bottom, Falcon®
: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
: Corning
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
Cell culture dishes, Falcon®
: Corning
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
: GLW
Round bottom high clarity polypropylene tube, disposable, Falcon®
: Corning
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF