ContactUsLinkComponent
HS Genomic DNA Diluent Marker, 120 mL. 1 bp lower marker. For use with the Fragment Analyzer systems and the DNF-467 kit. 1 * 120 mL
Catalog # AGLSDNF-375-0120
Supplier:  AGILENT
HS Genomic DNA Diluent Marker, 120 mL. 1 bp lower marker. For use with the Fragment Analyzer systems and the DNF-467 kit. 1 * 120 mL
Catalog # AGLSDNF-375-0120
Supplier:  AGILENT
Frequently Bought Together