Orders LinkContactUsLinkComponent
Bioanalyzer High Sensitivity Protein Analysis
Bioanalyzer High Sensitivity Protein Analysis
Agilent high sensitivity protein 250 kit, For sizing and highly sensitive quantitation of protein samples from 10 - 250 kDa, the kit includes 10 microfluidic chips, reagents and consumables, sufficient for 100 samples
Catalog # AGLS5067-1575
Supplier:   AGILENT
Bioanalyzer High Sensitivity Protein Analysis
Catalog # AGLS5067-1575
Supplier:  AGILENT
Bioanalyzer High Sensitivity Protein Analysis
Agilent high sensitivity protein 250 kit, For sizing and highly sensitive quantitation of protein samples from 10 - 250 kDa, the kit includes 10 microfluidic chips, reagents and consumables, sufficient for 100 samples
Price
SFr. 1’490.82
Log in for availability
1 * 1 ST
Product Details & Documents
High sensitivity electrophoretic separation of proteins and peptides up to 250 kD.
  • A ready-to-use system featuring standardised assays and pre-packaged reagent kits which are suitable to analyse samples up to 250 kD
  • After sample labelling, fast results are achieved with automated electrophoresis of 10 samples in 30 minutes
  • Highly sensitive with an accuracy down to silver stain levels, approximately 1 pg/µl of labelled protein on chip
  • Fully operational across a large linear dynamic range with four orders of magnitude
  • Highly precise impurity detection, down to 0,05% and lower
Perform high sensitivity protein electrophoresis with the 2100 Bioanalyzer system. The high sensitivity protein 250 assay provides a broad dynamic range with sensitivity down to silver stain comparable levels. Efficiency is enhanced by eliminating handling of SDS-PAGE slab gels, staining or, imaging steps.
The system provides a fast, automated, objective and flexible solution for protein and peptide characterisation, quality control, and impurity detection in the low picogram range. Compatible sample types include protein lysates, purified protein and peptides, antibodies, and protein stability tests.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    Agilent high sensitivity protein 250 kit
  • Seq:
    01
  • Details:
    For sizing and highly sensitive quantitation of protein samples from 10 - 250 kDa, the kit includes 10 microfluidic chips, reagents and consumables, sufficient for 100 samples