Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PLATE COUNT AGAR BOTTLED 10x500ML 1 * 5 L
Catalog # OXOIBO0195V
Supplier:
THERMO FISHER DIAGNOSTICS
Certificates
Expand image
PLATE COUNT AGAR BOTTLED 10x500ML 1 * 5 L
Catalog # OXOIBO0195V
Supplier:
THERMO FISHER DIAGNOSTICS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
VWR® Culture Media, Ready to Use, in Bottles
Supplier: VWR Chemicals
NUTRIENT AGAR ISO 16266 1 * 100 g
Supplier: LIOFILCHEM
Antibiotic Sensitivity Discs
Supplier: HIMEDIA
SRK® Surfaces and Equipment Microbial Testing Devices
Supplier: Copan
TEST IDENTIFICAZ./CLASSIFICA- ZIONE 1 * 1 items
Supplier: LIOFILCHEM
Membrane filters with dehydrated media pads for microbiological control
Supplier: Sartorius
Antibiotic assay discs
Supplier: LIOFILCHEM