Ca:4000mg/l K:4000mg/l Mg:600mg/l Mn:20mg/l Na:2000mg/l P:2000mg/l Zn:40 mg/l in HNO3 1 * 500 mL
Catalog # CMPLRM023977L5
Supplier: CUSTOM MADE CHEMICALS LAB
Ca:4000mg/l K:4000mg/l Mg:600mg/l Mn:20mg/l Na:2000mg/l P:2000mg/l Zn:40 mg/l in HNO3 1 * 500 mL
Catalog # CMPLRM023977L5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Organic reference standard, spinetoram, Dr Ehrenstorfer
Supplier: EHRENSTORFER
AMIODARONE RELATED COMPOUND E 1 * 100 mg
Supplier: Merck
ANTIMONY - SB @ 1000 µG/G IN HYDROCARBON OIL 1 * 1 items
Supplier: LGC Standards PROMOCHEM
Pesticide reference standard, cis-Permethrin 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
COLOUR REFERENCE SOL. GY ACC. TO PH EUR, 1 * 1 KIT
Supplier: Merck
4,4´-DDE 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
Methyl Linolenate, Reference Standard
Supplier: USP
Copper edetate solution Reag. Ph. Eur. 1022300
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Dichlorvos (2,2-dichlorovinyl dimethyl phosphate, DDVP)
Supplier: EHRENSTORFER
Alpha Tocopheryl Acid Succinate, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, Geraniol solution
Supplier: Merck
BUFFER SOLUTION PH1.68 / 0.01, 25C 1 * 500 mL
Supplier: REAGECON