ContactUsLinkComponent
4 components: Fluorides (F-) 3mg/l Chlorides (Cl-) 500mg/l Nitrates (NO3-) 100mg/l Sulphates (SO42-) 500mg/l in Water CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at tem 1 * 250 mL
Catalog # CMPLRM087462L25
4 components: Fluorides (F-) 3mg/l Chlorides (Cl-) 500mg/l Nitrates (NO3-) 100mg/l Sulphates (SO42-) 500mg/l in Water CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Store under normal laboratory conditions, at tem 1 * 250 mL
Catalog # CMPLRM087462L25
Frequently Bought Together