ContactUsLinkComponent
,,11 components: 100mg/l each of Ag Al As Cd Cu Cr Fe Ni Pb Sn Zn in HNO3 5% HF 0.2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM071866L1
,,11 components: 100mg/l each of Ag Al As Cd Cu Cr Fe Ni Pb Sn Zn in HNO3 5% HF 0.2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months. (12 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM071866L1
Frequently Bought Together