Safety Cap II GL45, Screw cap GL45 made of PP-black, insert made of PTFE with 2 capillary connections (UNF 1/4 ) and 1 connection fo r air valve (UNF 1/4 ). Including 2 fittings for tubing O.D. 3,2 mm (PFA-black), 1 air valve with flag with note for 1 * 1 items
Catalog # 590-0027
Supplier: VWR Collection
Safety Cap II GL45, Screw cap GL45 made of PP-black, insert made of PTFE with 2 capillary connections (UNF 1/4 ) and 1 connection fo r air valve (UNF 1/4 ). Including 2 fittings for tubing O.D. 3,2 mm (PFA-black), 1 air valve with flag with note for 1 * 1 items
Catalog # 590-0027
Supplier: VWR Collection
Frequently Bought Together
VWR®, Fittings for Capillary Connector
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Air Valve
Supplier: VWR Collection
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for titrators
Supplier: Mettler - Toledo
Supelco® Organic Reference Standard, Oenothein B
Supplier: Merck
Safety Waste Caps, S50
Supplier: S.C.A.T.