Safety Cap I GL45, Screw cap GL45 made of PP-black, insert made of PTFE with 1 capillary connection (UNF 1/4 ) and 1 connection for air valve (UNF 1/4 ). Including 1 fitting for tubing O.D. 3,2 mm (PFA-black), 1 air valve with flag with note for exc 1 * 1 items
Catalog # 590-0026
Supplier: VWR Collection
Safety Cap I GL45, Screw cap GL45 made of PP-black, insert made of PTFE with 1 capillary connection (UNF 1/4 ) and 1 connection for air valve (UNF 1/4 ). Including 1 fitting for tubing O.D. 3,2 mm (PFA-black), 1 air valve with flag with note for exc 1 * 1 items
Catalog # 590-0026
Supplier: VWR Collection
Frequently Bought Together
VWR®, Fittings for Capillary Connector
Supplier: VWR Collection
Consumables for Shimadzu autosamplers
Supplier: Trajan Scientific and Medical
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Centrifuge tubes
Supplier: Brand
Pasteur pipettes, disposable
Supplier: Brand
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
Laboratory bottles, narrow neck, with thread, amber glass, DURAN®
Supplier: DWK Life Sciences
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Reducing tubing connectors, b.safe
Supplier: Bohlender
VWR®, Accessories for VWR® Filtration Apparatus
Supplier: VWR Collection
VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
VWR®, Safety Waste Caps
Supplier: VWR Collection
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
Cole Parmer® Dri-Block® BH-250 Heaters
Supplier: Cole-Parmer
Waste caps, b.safe
Supplier: Bohlender