1-METHYL-3-(TRIFLUOROMETHYL)PYRAZOLE-5-B 1 * 250 mg
Catalog # APOSPC412531-250MG
Supplier: Apollo Scientific
1-METHYL-3-(TRIFLUOROMETHYL)PYRAZOLE-5-B 1 * 250 mg
Catalog # APOSPC412531-250MG
Supplier: Apollo Scientific
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diethylene glycol monobutyl ether, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Acetonitrile 99.8% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific