Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Ethanolamine 99%

Bottle, 100 g
Catalog # APOSOR921532-100G
Corrosive
Health
Irritant
Supplier:   Apollo Scientific
Ethanolamine 99%
Catalog # APOSOR921532-100G
Supplier:  Apollo Scientific
CAS Number:  141-43-5
Ethanolamine 99%

Bottle, 100 g

CAS Number:  141-43-5
Price
SFr. 26.00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Boiling Pt:170 °C
  • Melting Pt:10…11 °C
  • Melting Pt:10 - 11 °C
  • Density:1.012
  • Flash Pt:91 °C
  • UN:2491
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications