ETHYL 2,5-DICHLOROBENZOATE 1 * 25 g
Catalog # APOSOR4529-25G
Supplier: Apollo Scientific
Frequently Bought Together
Iodomethane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.4% ACS
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
N-HEPTANE 95% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals