Caproic acid 99%
Bottle, 25 g
Catalog # APOSOR6007-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Hexanoic acid
- CAS Number:142-62-1
- Formula:C₆H₁₂O₂
- MDL Number:MFCD00004421
- Molecular Weight:116.16 g/mol
- Boiling Pt:202…203 °C
- Melting Pt:–3 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



