TRIPHENYLPHOSPHINE SULPHIDE 1 * 25 g
Catalog # APOSOR25786-25G
Supplier: Apollo Scientific
Frequently Bought Together
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals