4-OXA-6,6,7,7-TETRAFLUOROHEPTANE-1,2-DIO 1 * 5 g
Catalog # APOSPC6465-5G
Supplier: Apollo Scientific
4-OXA-6,6,7,7-TETRAFLUOROHEPTANE-1,2-DIO 1 * 5 g
Catalog # APOSPC6465-5G
Supplier: Apollo Scientific
Frequently Bought Together
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Dimethyl sulphoxide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
o-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck
Ethylenediamine 99+%
Supplier: Thermo Fisher Scientific
1,1,1,3,3,3-Hexafluoro-2-propanol 99.5+%, pure
Supplier: Thermo Fisher Scientific
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals