4-OXA-6,6,7,7-TETRAFLUOROHEPTANE-1,2-DIO 1 * 1 g
Catalog # APOSPC6465-1G
Supplier: Apollo Scientific
4-OXA-6,6,7,7-TETRAFLUOROHEPTANE-1,2-DIO 1 * 1 g
Catalog # APOSPC6465-1G
Supplier: Apollo Scientific
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Cyclohexane ≥99.7%, SPECTRONORM® for UV spectrophotometry
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% for peptide synthesis
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Cyclohexane, EMPLURA®, Supelco®
Supplier: Merck
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals