4-OXA-6,6,7,7-TETRAFLUOROHEPTANE-1,2-DIO 1 * 250 mg
APOSPC6465-250MG
Frequently Bought Together
1,2,4-Trichlorobenzene ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Tetrahydrofuran ≥99% stabilised
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99%
: Thermo Fisher Scientific
tert-Butyl methyl ether for HPLC
: Thermo Fisher Scientific
Cyclohexane, extra dry 99.5%, AcroSeal®
: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
Water ACS, Deionized
: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
: Thermo Fisher Scientific
Diisopropyl ether 98+% stabilised, pure
: Thermo Fisher Scientific
Amyl Alcohol according to NF V 04-210 for analysis 1 * 1.000 mL
: PanReac AppliChem