7-BROMO-2,3-DIHYDROTHIENO[3,4-B][1,4]DIO 1 * 250 mg
Catalog # APOSOR8636-250MG
Supplier: Apollo Scientific
7-BROMO-2,3-DIHYDROTHIENO[3,4-B][1,4]DIO 1 * 250 mg
Catalog # APOSOR8636-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals