2-BROMO-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIO 1 * 1 g
Catalog # APOSOR71046-1G
Supplier: Apollo Scientific
2-BROMO-5-(4,4,5,5-TETRAMETHYL-1,3,2-DIO 1 * 1 g
Catalog # APOSOR71046-1G
Supplier: Apollo Scientific
Frequently Bought Together
Benzyl alcohol ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
ASSAY VOLLSTÄNDIGE ZELLE ABLÖSUNG REAGEN
Supplier: DISCOVERX
Glycerine
Supplier: Roth Carl
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane ≥99.5%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals