ContactUsLinkComponent
Masterflex® Nylon Clamp for Sanitary Fittings, 2"; 10/PK
Masterflex® Nylon Clamp for Sanitary Fittings, 2"; 10/PK
Catalog # MFLX12805-05
undefined
Masterflex® Nylon Clamp for Sanitary Fittings, 2"; 10/PK
Catalog # MFLX12805-05
  • Sanitary clamp size (port 1):
  • 2"
  • 1/2"
  • 1-1/2"
  • Sterilisation:
  • Autoclave, Gamma
  • Autoclave, Gamma
  • Autoclave, Gamma
  • Currently Viewing
Specifications
About this item
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
  • Description:
    Nylon Clamp for Sanitary Fittings, 2"; 10/PK
  • Material:
    Nylon
  • Seq:
    03
  • Environmentally Preferable:
  • Sanitary clamp size (port 1):
    2"
  • Sterilisation:
    Autoclave, Gamma
  • Pk:
    10
  • Clamp type:
    Sanitary
Frequently Bought Together