TERT-BUTYL N-[2-(1,2,3,4-TETRAHYDROQUINOLIN-8-YL)ETHYL]CARBAMATE 1 * 1 g
Catalog # UKROBBV41716700-1G
Supplier: UKRORGSYNTHESIS
TERT-BUTYL N-[2-(1,2,3,4-TETRAHYDROQUINOLIN-8-YL)ETHYL]CARBAMATE 1 * 1 g
Catalog # UKROBBV41716700-1G
Supplier: UKRORGSYNTHESIS
Frequently Bought Together
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals