AMBINTER Deoxysappanone B (3R)-3-[(3,4-dihydroxyphenyl)methyl]-7-hydroxy-2,3-dihydrochromen-4-one, CAS 113122-54-6 1 * 10 mg
MSPPAMB22172794
AMBINTER Deoxysappanone B (3R)-3-[(3,4-dihydroxyphenyl)methyl]-7-hydroxy-2,3-dihydrochromen-4-one, CAS 113122-54-6 1 * 10 mg
MSPPAMB22172794
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals