AMBINTER (3R)-3-hydroxy-1-(4-hydroxyphenyl)-5-phenylpentan-1-one, CAS 54835-64-2 1 * 10 mg
Catalog # MSPPAMB22173713
Supplier: MARKET SOURCE PART PROCESS
AMBINTER (3R)-3-hydroxy-1-(4-hydroxyphenyl)-5-phenylpentan-1-one, CAS 54835-64-2 1 * 10 mg
Catalog # MSPPAMB22173713
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99.9% stabilised, PESTINORM® for MOSH/MOAH analysis
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Trichloroethylene 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals