LM- HYPURE WFI QUAL WATER, 100L BODY MPC 1 * 100 L
Catalog # SH30221.51
Supplier: HyClone products (Cytiva)
LM- HYPURE WFI QUAL WATER, 100L BODY MPC 1 * 100 L
Catalog # SH30221.51
Supplier: HyClone products (Cytiva)
Frequently Bought Together
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Cryoboxes, 133×133 mm
Supplier: TENAK
MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Freezer boxes (Cryoboxes), True North®
Supplier: HEATHROW
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Cryo.S™ tubes, 1 and 2 ml
Supplier: Greiner Bio-One
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Supplier: EPPENDORF
TruCool® Ice Pans with Lids, Multi-Purpose Containers
Supplier: BIOCISION
Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
Sample tubes for stool, with screw cap
Supplier: BOETTGER PAUL
PARTITION INSERTS FOR CRYOBOX 10X10 1 * 10 items
Supplier: Roth Carl
Eppendorf® Assay/Reader Microplates
Supplier: EPPENDORF
Cryogenic vials, Cryovial®
Supplier: Simport Scientific