ContactUsLinkComponent
LONG FINE FOCUS X-RAY TUBE (C-tech) Target: Cu, 2200 W max., Focal spot size: 0.4 x 12 mm Required cooling water: 3.5 l/ min at 4,5 bar 1 * 1 ST
  MSPP9001004
LONG FINE FOCUS X-RAY TUBE (C-tech) Target: Cu, 2200 W max., Focal spot size: 0.4 x 12 mm Required cooling water: 3.5 l/ min at 4,5 bar 1 * 1 ST
  MSPP9001004
Frequently Bought Together