2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile,97% 1 * 1 g
Catalog # AMBDA505090-1G
Supplier: AMBEED
2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile,97% 1 * 1 g
Catalog # AMBDA505090-1G
Supplier: AMBEED
Frequently Bought Together
ASSAY VOLLSTÄNDIGE ZELLE ABLÖSUNG REAGEN
Supplier: DISCOVERX
2,2,4-Trimethylpentane ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene, extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
DICHLOROMETHANE EXTRA PURE. DAB. STABILI 1 * 100 L
Supplier: Honeywell Chemicals
Diethyl Ether stabilized with &sim 6 ppm of BHT (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1.000 mL
Supplier: PanReac AppliChem
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific