2-Amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol,95% 1 * 0,25 g
Catalog # AMBDA529957-250MG
Supplier: AMBEED
2-Amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol,95% 1 * 0,25 g
Catalog # AMBDA529957-250MG
Supplier: AMBEED
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals