N-Butyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine,98% 1 * 0,25 g
Catalog # AMBDA553198-250MG
Supplier: AMBEED
N-Butyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine,98% 1 * 0,25 g
Catalog # AMBDA553198-250MG
Supplier: AMBEED
Frequently Bought Together
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
n-Hexane ≥95% for spectrophotometry
Supplier: Thermo Fisher Scientific
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% (by GC) for UV and IR spectrophotometry, for HPLC
Supplier: PanReac AppliChem
N-Ethyldiisopropylamine 99%
Supplier: Thermo Fisher Scientific
Cyclopentyl methyl ether (CPME) ≥99.9%
Supplier: Thermo Fisher Scientific
Acetone 99.6% ACS
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific