4-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile,97% 1 * 0,1 g
Catalog # AMBDA600438-100MG
Supplier: AMBEED
4-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile,97% 1 * 0,1 g
Catalog # AMBDA600438-100MG
Supplier: AMBEED
Frequently Bought Together
Cyclohexane anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Dimethyl sulphoxide, Bioreagent
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Heptafluorobutyric anhydride (HFAA), Supelco®
Supplier: Merck
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific