SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
N,N-Bis(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)aniline,95% 1 * 0,1 g
AMBDA1529434-100MG
:
AMBEED
N,N-Bis(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)aniline,95% 1 * 0,1 g
AMBDA1529434-100MG
:
AMBEED
Frequently Bought Together
Glycerine
: Roth Carl
Pyridine-3-sulphonyl chloride ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
: Merck
2-Butoxyethanol 99%, extra pure
: Thermo Fisher Scientific
1-Propanol ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
Water ACS
: Thermo Fisher Scientific
Pyridine ≥99%
: Thermo Fisher Scientific
Tetrahydrofuran 99+% stabilised ACS
: Thermo Fisher Scientific
tert-Butyl methyl ether for analysis
: Thermo Fisher Scientific
Liquid for manometer with sloping tube
: VWR Chemicals
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
: MACRON AVANTOR BRAND