1-Ethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole,95% 1 * 0,25 g
Catalog # AMBDA690919-250MG
Supplier: AMBEED
1-Ethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole,95% 1 * 0,25 g
Catalog # AMBDA690919-250MG
Supplier: AMBEED
Frequently Bought Together
Acetone, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dipentyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, extra dry 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Oxidation-reduction (ORP) standards solutions, ZoBell's
Supplier: Hach
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
Glycerine ≥86%
Supplier: Roth Carl
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals