2-Chloro-4-methylbenzoic acid,97% 1 * 10 g
Catalog # AMBDA158504-10G
Supplier: AMBEED
Frequently Bought Together
Dipentyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Methanol, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) EMPARTA® ACS analytical reagent
Supplier: Merck
N,N-Dimethylhydrazine
Supplier: Merck
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
4-(Methylamino)phenol sulphate 99.0-101.5% ACS
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Chloroform stabilized with ethanol pure 1 * 1.000 mL
Supplier: PanReac AppliChem
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Triethylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals